Sigma-Aldrich
IL-22 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
IL-22 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sign Into View Organizational & Contract Pricing Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
IL-22 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sign Into View Organizational & Contract Pricing