MilliporeSigma
  • Gemcitabine loaded biodegradable PLGA nanospheres for in vitro pancreatic cancer therapy.
Gemcitabine loaded biodegradable PLGA nanospheres for in vitro pancreatic cancer therapy.
Materials science & engineering. C, Materials for biological applications (2014-12-11)
L R Jaidev, Uma Maheswari Krishnan, Swaminathan Sethuraman
ABSTRACT
Pancreatic cancer is the fourth leading cancer with 85% mortality rate in USA alone and it is prevalent in many other developed and developing countries. Clinically, gemcitabine is prescribed as the first line chemotherapeutic drug for pancreatic cancer treatment. Gemcitabine-loaded poly(lactide-co-glycolide) (PLGA) nanospheres were synthesized and their physico-chemical properties were evaluated. The FESEM images showed that the gemcitabine loaded and blank nanospheres were 180 nm and 200 nm, respectively. The optimized encapsulation efficiency of gemcitabine was 15%. It was observed that 100% of gemcitabine was released from the PLGA nanospheres for 41 days in phosphate buffered saline (PBS) at pH7.4. The uptake of nanospheres in MiaPaCa-2 cells was studied using sulforhodamine B loaded PLGA nanospheres and our results showed that the nanospheres were taken up within 3h. Furthermore, the cytotoxicity of PLGA nanospheres loaded with gemcitabine showed a relative decrease in IC50 in MiaPaCa-2 and ASPC-1 pancreatic cancer cells in comparison to free gemcitabine. The study demonstrates that this system hold promise to improve the therapeutic efficacy of gemcitabine in vitro.
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Chloroform, suitable for HPLC
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer