MilliporeSigma
  • Anaphylaxis induced by a drug containing lysozyme and papain: influence of papain on the IgE response.
Anaphylaxis induced by a drug containing lysozyme and papain: influence of papain on the IgE response.
International archives of allergy and immunology (2014-11-02)
Sara Benedé, Inmaculada Pérez-Rangel, Daniel Lozano-Ojalvo, Elena Molina, María Dolores Ibañez, Rosina López-Fandiño, Iván López-Expósito
ABSTRACT
This paper reports the case of an egg-allergic pediatric patient who, once desensitized to egg following a successful rush oral immunotherapy protocol, could also tolerate Lizipaina®, a drug containing lysozyme (LYS) and papain, which had previously caused him a severe allergic reaction. Because the LYS amount that elicited the anaphylactic reaction (5 mg) was much lower than that tolerated during a double-blind placebo-controlled food challenge (corresponding to approximately 60 mg of LYS), the possibility that the presence of papain could increase the allergenic potential of LYS was investigated. Lizipaina, LYS and LYS hydrolyzed with papain were analyzed by SDS-PAGE under reducing and nonreducing conditions, and Western blotting of sera from egg-allergic patients was performed in order to detect IgE-binding fragments. Finally, sequence identification of the IgE-reactive bands was carried out by MALDI-TOF/TOF. The SDS-PAGE pattern of LYS treated with papain under nonreducing conditions showed the presence of intact LYS that partially disappeared following reduction with β-mercaptoethanol, releasing IgE-reactive fragments as determined by Western blotting. MALDI-TOF/TOF revealed that papain degraded LYS, giving rise to three IgE-binding fragments: LYS (22-129), LYS (34-96) and LYS (62-128) that likely remained linked through the disulfide bonds present in the LYS molecule. The combined administration of LYS with proteolytic enzymes such as papain may have developed a severe allergic reaction in the patient studied, underlining the importance of considering all the components and their interactions when drugs are to be consumed by allergic persons.
Product Description
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
SAFC
Tricine
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%