Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)
Sign Into View Organizational & Contract Pricing Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sorbitan monopalmitate
Sign Into View Organizational & Contract Pricing Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, 99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sign Into View Organizational & Contract Pricing Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sign Into View Organizational & Contract Pricing USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sign Into View Organizational & Contract Pricing