Sigma-Aldrich
Chloroform, suitable for HPLC
Sign Into View Organizational & Contract Pricing Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Supelco
Chloroform, analytical standard
Sign Into View Organizational & Contract Pricing Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sign Into View Organizational & Contract Pricing