MilliporeSigma
  • Erbium-yttrium-aluminum-garnet laser irradiation ameliorates skin permeation and follicular delivery of antialopecia drugs.
Erbium-yttrium-aluminum-garnet laser irradiation ameliorates skin permeation and follicular delivery of antialopecia drugs.
Journal of pharmaceutical sciences (2014-09-05)
Woan-Ruoh Lee, Shing-Chuan Shen, Ibrahim A Aljuffali, Yi-Ching Li, Jia-You Fang
ABSTRACT
Alopecia usually cannot be cured because of the available drug therapy being unsatisfactory. To improve the efficiency of treatment, erbium-yttrium-aluminum-garnet (Er-YAG) laser treatment was conducted to facilitate skin permeation of antialopecia drugs such as minoxidil (MXD), diphencyprone (DPCP), and peptide. In vitro and in vivo percutaneous absorption experiments were carried out by using nude mouse skin and porcine skin as permeation barriers. Fluorescence and confocal microscopies were used to visualize distribution of permeants within the skin. Laser ablation at a depth of 6 and 10 μm enhanced MXD skin accumulation twofold to ninefold depending on the skin barriers selected. DPCP absorption showed less enhancement by laser irradiation as compared with MXD. An ablation depth of 10 μm could increase the peptide flux from zero to 4.99 and 0.33 μg cm(-2) h(-1) for nude mouse skin and porcine skin, respectively. The laser treatment also promoted drug uptake in the hair follicles, with DPCP demonstrating the greatest enhancement (sixfold compared with the control). The imaging of skin examined by microscopies provided evidence of follicular and intercellular delivery assisted by the Er-YAG laser. Besides the ablative effect of removing the stratum corneum, the laser may interact with sebum to break up the barrier function, increasing the skin delivery of antialopecia drugs. The minimally invasive, well-controlled approach of laser-mediated drug permeation offers a potential way to treat alopecia. This study's findings provide the basis for the first report on laser-assisted delivery of antialopecia drugs.
Product Description
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Rhodamine B, for fluorescence
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
Rhodamine B, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Ethyl 2-cyanoacrylate, liquid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
1,2-Propanediol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 ea
In Stock
Details...
$1,820.00