MilliporeSigma
mAbs Downstream
Products for Development and Manufacturing
Higher cell densities and antibody titers from increasingly advanced upstream processes result in complex downstream purification challenges. The focus of downstream bioprocessing is efficient recovery and purification while controlling bioburden and assuring viral safety to provide drug safety for patients. The downstream workflow, which includes clarification, chromatography, concentration, viral clearance, and sterile filtration steps, is essential for ensuring monoclonal antibody (mAb) product quality, yield, safety, and sterility. 
Polishing Chromatography
Ion exchange chromatography (IEC or IEX) is used in downstream processing to further purify monoclonal antibodies (mAbs) from product and process-related impurities, and viruses. Cation exchange (CEX) chromatography removes DNA, endotoxins, host cell proteins (HCPs), and leached Protein A ligands from the mAb feedstream. This step can also help reduce mAb aggregates. Anion exchange (AEX) chromatography removes trace contaminants such as nucleic acids and HCPs and can also contribute to virus clearance. Besides cation and anion exchangers, hydrophobic interaction chromatography (HIC) or mixed mode/multimodal chromatography can also be deployed in this polishing step. To optimize chromatographic separation and quickly evaluate media performance, high throughput screening (HTS) using pre-packed mini or robotic columns can be used. Both, resin (conventional beads) and membrane (adsorbers) formats, can be used for these applications.
For these polishing steps, we offer the following solutions:
Related Products
Product No.Product NameDescriptionPricing
AB152Anti-Tyrosine Hydroxylase AntibodyChemicon®, from rabbit
MAB377Anti-NeuN Antibody, clone A60clone A60, Chemicon®, from mouse
AB9610Anti-Olig-2 AntibodyChemicon®, from rabbit
MABE343Anti-Puromycin Antibody, clone 12D10clone 12D10, from mouse
AB5320Anti-NG2 Chondroitin Sulfate Proteoglycan AntibodyChemicon®, from rabbit
05-321Anti-Phosphotyrosine Antibody, clone 4G10®clone 4G10®, Upstate®, from mouse
MAB1281Anti-Nuclei Antibody, clone 235-1clone 235-1, Chemicon®, from mouse
07-449Anti-trimethyl-Histone H3 (Lys27) AntibodyUpstate®, from rabbit
MAB374Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5clone 6C5, Chemicon®, from mouse
MAB318Anti-Tyrosine Hydroxylase Antibody, clone LNC1ascites fluid, clone LNC1, Chemicon®
A1978Anti-β-Actin antibody, Mouse monoclonalclone AC-15, purified from hybridoma cell culture
AB5541Anti-Glial Fibrillary Acidic Protein AntibodyChemicon®, from chicken
ABN78Anti-NeuN Antibody (rabbit)from rabbit, purified by affinity chromatography
A0170Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goataffinity isolated antibody
MAB3420Anti-Tau-1 Antibody, clone PC1C6clone PC1C6, Chemicon®, from mouse
A5441Monoclonal Anti-β-Actin antibody produced in mouseclone AC-15, ascites fluid
V9131Monoclonal Anti-Vinculin antibody produced in mouseclone hVIN-1, ascites fluid
MAB1501Anti-Actin Antibody, clone C4ascites fluid, clone C4, Chemicon®
MAB2166Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8ascites fluid, clone 1HU-4C8, Chemicon®
MAB360Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5ascites fluid, clone GA5, Chemicon®
Related Categories
Four containers are depicted in the image related to buffers and pH adjusters. The two smaller containers on the left have a red lid and white body. The two larger containers on the right are blue with red-rimmed lids.
Buffers & pH Adjusters
Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs.
Learn More
An industrial clarification filter system featuring multiple cylindrical units mounted on a metal frame, complete with gauges and valves, designed for large-scale filtration processes.
Clarification & Harvest
Clarification steps prepare your bioreactor harvest material for downstream purification by reducing the levels of impurities and particulates.
Learn More
Chromatograph system for bioprocessing, featuring various tubes, dials, and components, predominantly silver with blue accents and a logo featuring the letter ‘M’ on the side panel.
Chromatography
Downstream purification in bioprocessing includes one or more chromatographic separation steps to purify your molecule and remove impurities.
Learn More
A female scientist wearing protective goggles holds up a sterile drug product.
Sterile Filtration and Bioburden Control
Our broad portfolio of membrane filters offers options for efficient processing of every pharma and biopharma stream.
Learn More
Dropper transferring liquid in monoclonal antibody process
Viral Clearance
Our viral clearance portfolio includes the virus filtration solutions, chemicals, holders and systems.
Learn More
Mobius® FlexReady Solution for TFF
Ultrafiltration & Diafiltration
Ultrafiltration & diafiltration increase capacity, concentration, and recovery in bioprocessing.
Learn More
Related Resources
Explore Our Solutions